missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ DS-33 GeneScan™ Install Kit Dx with GeneScan™ 600 LIZ™ Size Standard v2.0 Dx

Catalog No. A25793
Encompass
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A25793 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A25793 Supplier Applied Biosystems™ Supplier No. A25793

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Includes

Contents 4 tubes of GeneScan Installation Standard DS-33, 1 tube of GeneScan 600 LIZ size standard v2.0 Dx

Developed for use with Applied Biosystems fluorescence-based DNA electrophoresis systems. The use of internal size standard enables automated data analysis and is also essential for achieving high run to run precision in sizing DNA fragments.

The DS-33 GeneScan Install Kit Dx with GeneScan 600 LIZ Size Standard v2.0 Dx consists of pooled PCR products labeled with 6-FAM, VIC, NED, and PET dyes. To generate the pooled products, Human DNA is amplified with eight fluorescent PCR primer pairs, which amplify selected microsatellite loci (as listed in the package product insert). The kit also contains a tube of GeneScan 600 LIZ Size Standard v2.0 Dx (Cat. No. A25794).

The kit also contains a tube of GeneScan 600 LIZ™ size standard v2.0 Dx.

Order Info

1 Kit

Specifications

Content And Storage Each kit contains four tubes of GeneScan Installation Standard DS-33 and one tube of GeneScan 600 LIZ size standard v2.0 Dx.

Store the kit at -15 to -25°C until ready to use. Upon thawing, store the GeneScan 600 LIZ Size Standard v2.0 tube at 2°C to 8°C.
Detection Method Fluorescence
Format Tube
For Use With (Equipment) 3500 Dx Genetic Analyzer, SeqStudio Flex Dx Series
GC-Rich PCR Performance High
For Use With (Application) Sequencing
Label or Dye LIZ
Product Line GeneScan, LIZ
Product Type Installation Kit
Quantity 1 Kit
Shipping Condition Dry Ice
Test Time Fast
Show More Show Less

For In Vitro Diagnostic Use Only