missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Dry-Ready™ RT-LAMP Kit

Catalog No. A40006907
Change view
Click to view available options
Quantity:
200 Reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A40006907 200 Reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A40006907 Supplier Invitrogen™ Supplier No. A40006907

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Experience the simplicity and efficiency of the air drying process by gently heating materials at moderate temperatures in a simple ventilated oven.

The Dry-Ready RT-LAMP Kit is a complete solution for reverse transcription loop-mediated isothermal amplification (RT-LAMP)-based assay air drying. The kit includes all required components: Dry-Ready SuperScript IV Reverse Transcriptase, Dry-Ready Bst DNA Polymerase, Dry-Ready dNTP Mix, and 10X Dry-Ready RT-LAMP Reaction Buffer to facilitate pre- and post-drying procedures.

Features of the Dry-Ready RT-LAMP Kit include:

  • All required components for RT-LAMP assay drying and downstream applications
  • Rapid RNA conversion in RT-LAMP reaction using SuperScript IV Reverse Transcriptase
  • Robust Bst DNA Polymerase with enhanced speed and sensitivity, engineered for fast and efficient amplification in RT-LAMP

The Dry-Ready RT-LAMP Kit includes an evolved Bst DNA polymerase mutant with a fast reaction speed and increased sensitivity and tolerance to inhibitors, as well as SuperScript IV Reverse Transcriptase with increased processivity, thermostability, and inhibitor resistance.

The Dry-Ready kit is optimized for the air drying process. Notably, air drying is a simple process that does not impair functionality of the assay. On the contrary, it help provides flexibility on the initial volume of sample and increases sensitivity, resulting in a exceptional stabilization process for point-of-care applications.

Specifications

Product Type DNA Amplification Kit
Form Liquid
For Use With (Application) RT-LAMP, LAMP
Polymerase Bst DNA Polymerase
Quantity 200 Reactions
Research Category Nucleic Acid Amplification
Content And Storage

•Dry-Ready™ Bst DNA Polymerase
•Dry-Ready™ SuperScript™ IV Reverse Transcriptase
•Dry-Ready™ dNTP Mix
•5X Dry-Ready™ Reaction Buffer
•MgCl2 Solution
•Store at -15 to -25°C

No. of Reactions 200 Reactions
Optimal Reaction Temperature 65°C
Reverse Transcriptase SuperScript™ IV Reverse Transcriptase
Reaction Time ≤10 Min.
Show More Show Less

For Research Use Only.