missing translation for 'onlineSavingsMsg'
Learn More
Learn More
JUSTRITE Drum Storage Cabinets, 55 gal., Manual Close, with Drum Rollers
For storage of larger quantities of flammable solvents, the heavy-duty Drum Safety Cabinet is available in many styles and sizes.
$4,510.96 - $5,485.46
Specifications
| Certifications/Compliance | FM Approved |
|---|---|
| Depth (English) Exterior | 34 in. |
| Height (English) Exterior | 65 in. |
| Material | Steel (18 ga), Galvanized Steel |
| Product Type | Vertical Drum Storage Cabinet |
| Catalog Number | Mfr. No. | Color | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Color | Price | Quantity & Availability | |||||
|
18666744
|
JUSTRITE
896260* |
Yellow |
Each for $4,510.96
|
|
|||||
|
18666745
|
JUSTRITE
896261 |
Red |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
18666746
|
JUSTRITE
896262 |
Blue |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
18666747
|
JUSTRITE
896263* |
Gray |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
18666748
|
JUSTRITE
896265* |
White |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
Built to the same high standards as the smaller Classic Safety Cabinets. An array of drum accessories is available to ensure safety and convenience when storing and handling drums of hazardous materials.
- 18-gauge (1 mm) double-wall, welded steel with 38 mm of insulating air space for fire resistance.
- Half-depth shelf adjusts to accommodate a drum with pump or funnel, or stores spare drum accessories.
- Available in manual or self-close models.
- Choice of drum rollers to facilitate the movement of heavy drums or removable drum support to keep drums elevated off cabinet sump.
- Independently fire tested and approved by FM Approvals.
- High-performance, self-latching doors close easily and securely for maximum protection under fire conditions.
- U-Loc™ handle comes with a cylinder lock and key set or accepts an optional padlock.
- Haz-Alert™ reflective warning labels are highly visible under fire conditions.
- Patented SpillSlope™ galvanized steel shelves direct spills to back and bottom of a leakproof sump.
- Designed to meet OSHA and NFPA 30 standards. FM-approved and come with a ten-year warranty
Specifications
| FM Approved | |
| 65 in. | |
| Vertical Drum Storage Cabinet | |
| Manual-close, Locking, Three-point lock | |
| 350 lb./shelf | |
| 1 | |
| 346 lb. | |
| 30.8 in. | |
| 781 mm |
| 34 in. | |
| Steel (18 ga), Galvanized Steel | |
| 34 in. | |
| No | |
| 2 | |
| Powder-coat Paint | |
| 156.9 kg | |
| 864 mm |