missing translation for 'onlineSavingsMsg'
Learn More

JUSTRITE Drum Storage Cabinets, 55 gal., Manual Close, with Drum Rollers

Catalog No. 18666744
Change view
Click to view available options
Color:
Blue
Gray
Red
White
Yellow
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
18666744 Yellow
18666745 Red
18666746 Blue
18666747 Gray
18666748 White
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18666744 Supplier JUSTRITE Supplier No. 896260*

For storage of larger quantities of flammable solvents, the heavy-duty Drum Safety Cabinet is available in many styles and sizes.

Built to the same high standards as the smaller Classic Safety Cabinets. An array of drum accessories is available to ensure safety and convenience when storing and handling drums of hazardous materials.

  • 18-gauge (1 mm) double-wall, welded steel with 38 mm of insulating air space for fire resistance.
  • Half-depth shelf adjusts to accommodate a drum with pump or funnel, or stores spare drum accessories.
  • Available in manual or self-close models.
  • Choice of drum rollers to facilitate the movement of heavy drums or removable drum support to keep drums elevated off cabinet sump.
  • Independently fire tested and approved by FM Approvals.
  • High-performance, self-latching doors close easily and securely for maximum protection under fire conditions.
  • U-Loc™ handle comes with a cylinder lock and key set or accepts an optional padlock.
  • Haz-Alert™ reflective warning labels are highly visible under fire conditions.
  • Patented SpillSlope™ galvanized steel shelves direct spills to back and bottom of a leakproof sump.
  • Designed to meet OSHA and NFPA 30 standards. FM-approved and come with a ten-year warranty

Specifications

No. of Doors 2
Load Bearing Capacity 350 lb./shelf
Door Style Manual-close, Locking, Three-point lock
Weight (Metric) 156.9 kg
Width (English) Interior 30.8 in.
Width (Metric) Exterior 864 mm
Width (Metric) Interior 781 mm
No. of Shelves 1
Surface Coating Powder-coat Paint
Weight (English) 346 lb.
Explosion Relief Panel No
Width (English) Exterior 34 in.
Product Type Vertical Drum Storage Cabinet
Material Steel (18 ga), Galvanized Steel
Color Yellow
Certifications/Compliance FM Approved
Depth (English) Exterior 34 in.
Height (English) Exterior 65 in.
Show More Show Less