missing translation for 'onlineSavingsMsg'
Learn More

Eisco™ Human Melanoma

Catalog No. S30201
Change view
Click to view available options
Quantity:
1
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S30201 1
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S30201 Supplier Eisco™ Supplier No. BS18090

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Eisco Human Melanoma microscope slide containing a stained portion of human skin infected with melanoma

  • Microscope slide of human skin section showing melanoma
  • Melanoma is a form of skin cancer that develops in melanocytes, pigment-containing cells of skin
  • Pre-cleaned glass slide with ground edges
  • Professionally stained samples for best visualization of characteristic structures
  • Great choice for biology classrooms exploring structure-function relationships as per NGSS standards
  • Human skin section including melanoma

Specifications

Biosafety Level 1
Concentration 10%
Color Clear
Material Glass
Mounting Type Mounting Medium
Type Microbiology Slide
For Use With (Application) Microscopy
Manufacturing Origin China
Media Type Formalin
Packaging Cardboard Mailer
Quantity 1
Species Human
Size 1 x 3 in.
Slide Type Prepared Microbiology Slide
Show More Show Less