missing translation for 'onlineSavingsMsg'
Learn More

Drosha Rabbit anti-Human, Polyclonal, Bethyl Laboratories

Catalog No. 501561233
Encompass
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
501561233 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 501561233 Supplier Bethyl Laboratories Inc Supplier No. A301886A

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt.

Members of the ribonuclease III superfamily of double-stranded RNA-specific endoribonucleases participate in diverse RNA maturation and decay pathways in eukaryotic and prokaryotic cells processing in the nucleus .

Specifications

Antigen Drosha
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.2 mg/ml
Conjugate Unconjugated
Formulation TBS with 0.1% BSA and 0.09% sodium azide
Gene DROSHA
Gene Accession No. Q9NRR4
Gene Alias OTTHUMP00000220910; OTTHUMP00000220911; OTTHUMP00000220912; OTTHUMP00000220913; RNase III; drosha, double-stranded RNA-specific endoribonuclease; drosha, ribonuclease type III; nuclear RNase III Drosha; p241; putative protein p241 which interacts with transcription factor Sp1; putative ribonuclease III; ribonuclease 3; ribonuclease III, nuclear
Gene Symbols DROSHA
Host Species Rabbit
Immunogen Between 1324 and 1374
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 29102
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less