missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Eisco™ Dropping Funnel with Glass Key Stopcock
High quality borosilicate 3.3 glass cylindrical dropping funnels with glass key stopcocks and interchangeable polypropylene stoppers
Supplier: Eisco™ CH0472D
Description
Warranty
1 year limited manufacturer warranty
Chemical Identifiers
| Borosilicate Glass | |
| Clear | |
| funnel, stopper | |
| Glass key | |
| Cylindrical Dropping Funnel |
| Autoclavable | |
| Titration, neutralization | |
| 1 | |
| Polypropylene |
Specifications
| 500 mL | |
| 35.6 cm | |
| Clear | |
| funnel, stopper | |
| Glass key | |
| 29/32 |
| Borosilicate Glass | |
| Autoclavable | |
| Titration, neutralization | |
| 1 | |
| Polypropylene | |
| Cylindrical Dropping Funnel |