missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dronedarone Hydrochloride 98.0+%, TCI America™
$100.11 - $292.62
Chemical Identifiers
| CAS | 141625-93-6 |
|---|---|
| Molecular Formula | C31H45ClN2O5S |
| Molecular Weight (g/mol) | 593.22 |
| MDL Number | MFCD00914940 |
| InChI Key | DWKVCQXJYURSIQ-UHFFFAOYSA-N |
| Synonym | N-[2-Butyl-3-[4-[3-(dibutylamino)propoxy]benzoyl]-5-benzofuranyl]methanesulfonamide Hydrochloride |
| PubChem CID | 219025 |
| IUPAC Name | N-[2-butyl-3-[4-[3-(dibutylamino)propoxy]benzoyl]-1-benzofuran-5-yl]methanesulfonamide;hydrochloride |
| SMILES | CCCCC1=C(C2=C(O1)C=CC(=C2)NS(=O)(=O)C)C(=O)C3=CC=C(C=C3)OCCCN(CCCC)CCCC.Cl |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
D4689200MG
|
TCI America
D4689200MG |
200 mg |
Each for $100.11
|
|
|||||
|
D46891G
|
TCI America
D46891G |
1 g |
Each for $292.62
|
|
|||||
Chemical Identifiers
| 141625-93-6 | |
| 593.22 | |
| DWKVCQXJYURSIQ-UHFFFAOYSA-N | |
| 219025 | |
| CCCCC1=C(C2=C(O1)C=CC(=C2)NS(=O)(=O)C)C(=O)C3=CC=C(C=C3)OCCCN(CCCC)CCCC.Cl |
| C31H45ClN2O5S | |
| MFCD00914940 | |
| N-[2-Butyl-3-[4-[3-(dibutylamino)propoxy]benzoyl]-5-benzofuranyl]methanesulfonamide Hydrochloride | |
| N-[2-butyl-3-[4-[3-(dibutylamino)propoxy]benzoyl]-1-benzofuran-5-yl]methanesulfonamide;hydrochloride |
Specifications
| 141625-93-6 | |
| White | |
| MFCD00914940 | |
| N-[2-Butyl-3-[4-[3-(dibutylamino)propoxy]benzoyl]-5-benzofuranyl]methanesulfonamide Hydrochloride | |
| CCCCC1=C(C2=C(O1)C=CC(=C2)NS(=O)(=O)C)C(=O)C3=CC=C(C=C3)OCCCN(CCCC)CCCC.Cl | |
| 593.22 | |
| 593.22 | |
| Crystalline Powder |
| 144°C | |
| C31H45ClN2O5S | |
| 200 mg | |
| DWKVCQXJYURSIQ-UHFFFAOYSA-N | |
| N-[2-butyl-3-[4-[3-(dibutylamino)propoxy]benzoyl]-1-benzofuran-5-yl]methanesulfonamide;hydrochloride | |
| 219025 | |
| ≥98.0% (HPLC,T) | |
| Dronedarone Hydrochloride |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator