accessibility menu, dialog, popup

UserName

CAS RN 141625-93-6

IUPAC Name: hydrogen N-(2-butyl-3-{4-[3-(dibutylamino)propoxy]benzoyl}-1-benzofuran-5-yl)methanesulfonamide chloride
Synonyms: hydrogen multaq chloride
Molecular Weight (g/mol): 593.22
Molecular Formula: C31H45ClN2O5S
InChi Key: DWKVCQXJYURSIQ-UHFFFAOYSA-N
SMILES: [H+].[Cl-].CCCCN(CCCC)CCCOC1=CC=C(C=C1)C(=O)C1=C(CCCC)OC2=CC=C(NS(C)(=O)=O)C=C12
Percent Purity 
  • (2)
  • (1)
Physical Form 
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)