missing translation for 'onlineSavingsMsg'
Learn More

Dronedarone Hydrochloride 98.0+%, TCI America™

Catalog No. D4689200MG
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D4689200MG 200 mg
D46891G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D4689200MG Supplier TCI America Supplier No. D4689200MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 141625-93-6
Molecular Formula C31H45ClN2O5S
Molecular Weight (g/mol) 593.22
MDL Number MFCD00914940
InChI Key DWKVCQXJYURSIQ-UHFFFAOYSA-N
Synonym N-[2-Butyl-3-[4-[3-(dibutylamino)propoxy]benzoyl]-5-benzofuranyl]methanesulfonamide Hydrochloride
PubChem CID 219025
IUPAC Name N-[2-butyl-3-[4-[3-(dibutylamino)propoxy]benzoyl]-1-benzofuran-5-yl]methanesulfonamide;hydrochloride
SMILES CCCCC1=C(C2=C(O1)C=CC(=C2)NS(=O)(=O)C)C(=O)C3=CC=C(C=C3)OCCCN(CCCC)CCCC.Cl

Specifications

Melting Point 144°C
Color White
Quantity 200 mg
Formula Weight 593.22
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Dronedarone Hydrochloride
TSCA No
Recommended Storage Refrigerator