missing translation for 'onlineSavingsMsg'
Learn More

Drometrizole trisiloxane, ≥98% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111013241
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111013241 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111013241 Supplier Merck Emd Millipore Supplier No. 6879050MG

Drometrizole trisiloxane (DRT) belongs to the lipophilic benzotriazole derivative class of compounds commonly utilized in sunscreen cosmetics.

Featured Industries: Cleaning Products, Cosmetics, Environmental, Food and Beverages and Personal Care

Specifications

CAS 155633-54-8
Physical Form Neat
Linear Formula C24H39N3O3 Si3
MDL Number MFCD22373626
Synonym 2-(2 H-Benzotriazol-2-yl)-4-methyl-6-{2-methyl-3-{1,3,3,3-tetramethyl-1-[(trimethylsilyl)oxy]disiloxanyl}propyl}phenol; Silatrizole
Type Analytical Standard
Quantity 50 mg
Formula Weight 501.84
Percent Purity ≥98% (HPLC)
Grade Analytical Standard