missing translation for 'onlineSavingsMsg'
Learn More

Drierite™, without indicator, 20-40 mesh

Catalog No. p-45086
Change view
Click to view available options
Quantity:
454 g
2.3 kg
Packaging:
Glass Bottle
Plastic Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC219074540 454 g Glass Bottle
AC219070023 2.3 kg Plastic Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC219074540 Supplier Thermo Scientific Chemicals Supplier No. 219074540
Only null left

CAS: 7778-18-9 | CaO4S | 136.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Adsorbent, Indicator

Chemical Identifiers

CAS 7778-18-9
Molecular Formula CaO4S
Molecular Weight (g/mol) 136.13
MDL Number MFCD00010912
InChI Key OSGAYBCDTDRGGQ-UHFFFAOYSA-L
PubChem CID 24497
ChEBI CHEBI:31346
IUPAC Name calcium sulfate
SMILES [Ca++].[O-]S([O-])(=O)=O

Specifications

Melting Point 1450.0°C
Physical Form Granular
Quantity 454 g
Fieser 01,107
Merck Index 15, 1708
Packaging Glass Bottle
Solubility Information Solubility in water: ca. 0.25g/L (20°C). Other solubilities: insoluble in organic liquids, slightly soluble in glycerol. SiO2 and insolubles: 0.15%
R2O 0.14%
Formula Weight 136.14
Chemical Name or Material Drierite™, 20-40 mesh
Hazard Category
EINECSNumber 231-900-3
RTECSNumber WS6920000
TSCA TSCA

RUO – Research Use Only