missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Drierite™, without indicator, 10-20 mesh
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Adsorbent, Indicator
Specifications
Specifications
| Physical Form | Granular |
| Melting Point | 1450.0°C |
| Quantity | 500 g |
| Packaging | Glass Bottle |
| Solubility Information | Solubility in water: ca. 0.25g/L (20°C). Other solubilities: insoluble in organic liquids, slightly soluble in glycerol |
| IUPAC Name | calcium;sulfate |
| Formula Weight | 136.14 |
| Chemical Name or Material | Drierite™ |
Safety and Handling
| Hazard Category |
RUO – Research Use Only