missing translation for 'onlineSavingsMsg'
Learn More

Drierite™, regular (CaSO{4}), Lab Grade

Catalog No. AA89750P2
Change view
Click to view available options
Quantity:
1 lb.
5 lb.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA89750P2 1 lb.
AA89750P5 5 lb.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA89750P2 Supplier Thermo Scientific Chemicals Supplier No. 089750P2

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 7778-18-9 | CaO4S | 136.13 g/mol

Drierite™, regular (CaSO{4}), Lab Grade is used in drying of air, industrial gases, refrigerants and many organic liquids. It is used to maintain a dry atmosphere in storage spaces, vaults and commercial packages. It is utilized for the protection of hygroscopic materials, corrosion, rust, and other deterioration caused by high humidity. Further, it is used as an additive in polymer formulations.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7778-18-9
Molecular Formula CaO4S
Molecular Weight (g/mol) 136.13
MDL Number MFCD00010912
InChI Key OSGAYBCDTDRGGQ-UHFFFAOYSA-L
PubChem CID 24497
ChEBI CHEBI:31346
IUPAC Name calcium sulfate
SMILES [Ca++].[O-]S([O-])(=O)=O

Specifications

Physical Form Granular-8 Mesh
Color White
Density 2.96g/cm3
Material Calcium Sulfate
Quantity 1 lb.
Synonym Calcium sulfate, anhydrous
Solubility Information Slightly soluble in water.
Sensitivity Hygroscopic
Grade Laboratory
Chemical Name or Material Calcium Sulfate, Regular
EINECSNumber 231-900-3
RTECSNumber WS6920000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only