Learn More
Drierite™, with indicator, 4 mesh
CAS: 7778-18-9 | CaO4S | 136.13 g/mol
Supplier: Thermo Scientific Chemicals 219080020
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Adsorbent, IndicatorSpecifications
| 1450.0°C | |
| Blue | |
| 4 mesh | |
| Calcium Sulfate with Cobalt Chloride | |
| CaO4S | |
| 01,107 | |
| Glass Bottle | |
| OSGAYBCDTDRGGQ-UHFFFAOYSA-L | |
| calcium sulfate | |
| 24497 | |
| 136.13 |
| Irregular Granules or Chunks | |
| Drierite™ | |
| 7778-18-9,7646-79-9 | |
| 2 kg | |
| MFCD00010912 | |
| 15, 1708 | |
| Solubility in water: slightly soluble. Other solubilities: slightly soluble in glycerol, practically insoluble in most common organic, solvents | |
| [Ca++].[O-]S([O-])(=O)=O | |
| 136.13 | |
| CHEBI:31346 |
Chemical Identifiers
| 7778-18-9 | |
| 136.13 | |
| OSGAYBCDTDRGGQ-UHFFFAOYSA-L | |
| CHEBI:31346 |
| CaO4S | |
| MFCD00010912 | |
| 24497 | |
| [Ca++].[O-]S([O-])(=O)=O |
Safety and Handling
GHS H Statement:
May cause cancer by inhalation.
Suspected of causing genetic defects.
May damage fertility.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause an allergic skin reaction.
Toxic to aquatic life with long lasting effects.
GHS P Statement:
Do not breathe dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid release to the environment.
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
Use personal protective equipment as required.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
HYGROSCOPIC
EINECSNumber : 231-900-3
RTECSNumber : WS6920000
TSCA : TSCA
RUO – Research Use Only