missing translation for 'onlineSavingsMsg'
Learn More

Drierite™, indicating (CaSO{4})

CAS: 7778-18-9 | CaO4S | 136.13 g/mol

Supplier:  Thermo Scientific Chemicals 043063P5

 View more versions of this product

Catalog No. AA43063P5


Only null left
Explore available promotions

Description

Description

Drierite™, indicating (CaSO{4}) is used as a desiccant. It is used as a drying agent utilized for the efficient and rapid drying of air, industrial gases, refrigerants, organic liquids and solids. Further, it is used to maintain a dry atmosphere in storage spaces, vaults, commercial packages, and other enclosures. In addition to this, it is involved in the protection of hygroscopic materials.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Granular
Drierite
5 lb.
MFCD00010912
Soluble in water.
[Ca++].[O-]S([O-])(=O)=O
calcium sulfate
24497
Laboratory
Blue
7778-18-9
CaO4S
Odorless
OSGAYBCDTDRGGQ-UHFFFAOYSA-L
Hygroscopic
136.13
CHEBI:31346

Chemical Identifiers

7778-18-9
136.13
OSGAYBCDTDRGGQ-UHFFFAOYSA-L
CHEBI:31346
[Ca++].[O-]S([O-])(=O)=O
CaO4S
MFCD00010912
24497
calcium sulfate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

H334-H350-H317-H402-H412: May cause allergy or asthma symptoms or breathing difficulties if inhaled. May cause cancer. May cause an allergic skin reaction. Harmful to aquatic life. Harmful to aquatic life with long lasting effects.

EINECSNumber : 231-900-3

RTECSNumber : WS6920000

TSCA : Yes

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only