missing translation for 'onlineSavingsMsg'
Learn More

DRIERITE™, Indicating, 4 Mesh, J.T. Baker™

Catalog No. 2002758
Change view
Click to view available options
Quantity:
454 g
2.3 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
2002758 454 g
2002757 2.3 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 2002758 Supplier Avantor Supplier No. L05707

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7778-18-9
Molecular Formula CaO4S
Molecular Weight (g/mol) 136.13
MDL Number MFCD00010912
InChI Key OSGAYBCDTDRGGQ-UHFFFAOYSA-L
PubChem CID 24497
ChEBI CHEBI:31346
IUPAC Name calcium sulfate
SMILES [Ca++].[O-]S([O-])(=O)=O

Specifications

Quantity 454 g
Mesh Size 4 Mesh
Specific Gravity 1L = 1.87kg
Chemical Name or Material DRIERITE™, Indicating