missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DRIERITE™, Indicating, 4 Mesh, J.T. Baker™
Supplier: Avantor J.T.Baker L05702
Specifications
| DRIERITE™ | |
| 7778-18-9 | |
| CaO4S | |
| 4 Mesh | |
| [Ca++].[O-]S([O-])(=O)=O | |
| 136.13 | |
| CHEBI:31346 |
| Indicating | |
| 2.3 kg | |
| MFCD00010912 | |
| OSGAYBCDTDRGGQ-UHFFFAOYSA-L | |
| calcium sulfate | |
| 24497 | |
| 1L = 1.87kg |
Chemical Identifiers
| 7778-18-9 | |
| 136.13 | |
| OSGAYBCDTDRGGQ-UHFFFAOYSA-L | |
| CHEBI:31346 | |
| [Ca++].[O-]S([O-])(=O)=O |
| CaO4S | |
| MFCD00010912 | |
| 24497 | |
| calcium sulfate |