missing translation for 'onlineSavingsMsg'
Learn More

Drierite™ Absorbent, Indicating 10-20 Mesh, MilliporeSigma™

Catalog No. 361042002
Change view
Click to view available options
Quantity:
5 lb.
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
361042002 5 lb. Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 361042002 Supplier Technidata Supplier No. DX25193

Chemical Identifiers

CAS 7778-18-9 , 7646-79-9
Molecular Formula CaO4S
Molecular Weight (g/mol) 136.13
MDL Number MFCD00010912
InChI Key OSGAYBCDTDRGGQ-UHFFFAOYSA-L
Synonym calcium sulfate, drierite, gypsum, sulfuric acid, calcium salt 1:1, karstenite, calcium sulphate, anhydrous gypsum, plaster of paris, anhydrous calcium sulfate, calcium sulfate, anhydrous
PubChem CID 24497
ChEBI CHEBI:31346
IUPAC Name calcium sulfate
SMILES [Ca++].[O-]S([O-])(=O)=O

Specifications

Color Blue
CAS Max % Calcium sulfate: 90%
CAS Min % Cobalt(II) chloride: 5%
Physical Form Solid
Quantity 5 lb.
UN Number UN 3077
Odor No strong odor known
Packaging Glass bottle
Chemical Name or Material Calcium Sulphate, Cobaltous Chloride as indicator, 10-20 Mesh
Safety handling Check Alert storage code Red
Hazard Category H317 May cause an allergic skin reaction. H334 May cause allergy or asthma symptoms or breathing difficulties if inhaled. H341 Suspected of causing genetic defects
Hazard Statement P201 Obtain special instructions before use. P202 Do not handle until all safety precautions have been read and understood. P261 Avoid breathing dust/ fume/ gas/ mist/ vapors/ spray. P272 Contaminated work clothing must not be allowed out of the workplace. P280 Wear protective gloves. P302 + P352 IF ON SKIN: Wash with plenty of soap and water. P304 + P341 IF INHALED: If breathing is difficult, remove perso n to fresh air and keep comfortable for breathing. P308 + P313 IF exposed or concerned: Get medical advice/ attent ion. P321 Specific treatment (see supplemental first aid instructions on this label). P333 + P313 If skin irritation or rash occurs: Get medical advi ce/ attention. P363 Wash contaminated clothing before reuse. P405 Store locked up. P501 Dispose of contents/ container to an approved waste dispos al plant
DOTInformation 6.1 D Non combustible, acute toxicity cat. 3 / toxic or substances with chronic effects