missing translation for 'onlineSavingsMsg'
Learn More

Dried Ferrous Sulfate, Powder, USP, 86-89%, Spectrum™ Chemical

F1065, 13463-43-9, FeSO4·xH2O

Supplier:  Spectrum Chemical Mfg Cor F106512KG

 View more versions of this product

Catalog No. 18602355


Only null left
Explore available promotions

Description

Description

Spectrum™ Chemical Dried Ferrous Sulfate, Powder, USP is used to prevent or treat low levels of iron in the blood. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Specifications

Specifications

13463-43-9
12 kg
XBDUTCVQJHJTQZ-UHFFFAOYSA-L
λ²-iron(2+) hydrate sulfate
86 to 89%
Poly Pail
100%
FeH2O5S
O.[Fe++].[O-]S([O-])(=O)=O
169.92
USP

Chemical Identifiers

13463-43-9
169.92
λ²-iron(2+) hydrate sulfate
FeH2O5S
XBDUTCVQJHJTQZ-UHFFFAOYSA-L
O.[Fe++].[O-]S([O-])(=O)=O
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions