missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ dRhodamine Dye Terminator Ready Reaction Cycle Sequencing Kit

Catalog No. 403045
Encompass
Change view
Click to view available options
No. of Reactions:
100 Reactions
1000 Reactions
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Reactions
403045 1000 Reactions
403044 100 Reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 403045 Supplier Applied Biosystems™ Supplier No. 403045

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Includes

Each kit contains dRhodamine terminator ready reaction mix, M13 (-21) primer, pGEM control DNA, and a user's manual.

The dRhodamine Terminator Cycle Sequencing Kit provides optimized dRhodamine dideoxy terminator chemistries for decreased background noise in a wide array sequencing applications.

  • Increase your signal-to-noise ratio
  • Simplify workflows with easy-to-clean-up chemistries
  • Get optimized chemistry for sequencing templates with homo-polymers (AA's, TT's, etc.)
  • Obtain clean signal, even peak patterns, and minimal mobility shifts with improved dye terminators
  • Reduce spectrum overlap with narrow dye emission spectra

Specifications

Content And Storage Each kit contains dRhodamine Terminator Ready Reaction Mix, M13 (-21) Primer, pGEM Control DNA, and a user's manual. There are sufficient reagents for 1000 reactions. Store kit at -15 to -25°C and avoid repeated freeze-thaw cycles.
Format 8-well Strip, 96-well Plate
For Use With (Equipment) 310 Genetic Analyzer, 3130xl Genetic Analyzer, ABI PRISM™ 377, GeneAmp 2700, GeneAmp 9600, GeneAmp 9700, GeneAmp™ 2400, 3130 Genetic Analyzer
For Use With (Application) Standard Sequencing
Includes dRhodamine Dye Terminator Ready Reaction Cycle Sequencing Kit, 1000 rxns, 8-well Strips, 96-well Plate, For 310 Genetic Analyzer, 3130 Genetic Analyzer
No. of Reactions 1000 Reactions
Product Type Sequencing Kit
Quantity 1,000 reactions
Technique Fluorescent Dye Terminator Sequencing
Template Compatibility Single Stranded DNA, Plasmid DNA (≤15Kb), PCR Amplicons

For Research Use Only. Not for use in diagnostic procedures.