missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ dRhodamine Dye Terminator Ready Reaction Cycle Sequencing Kit

Catalog No. 403044
Encompass
Change view
Click to view available options
No. of Reactions:
100 Reactions
1000 Reactions
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Reactions
403044 100 Reactions
403045 1000 Reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 403044 Supplier Applied Biosystems™ Supplier No. 403044

Includes

Each kit contains 800uL tube of dRhodamine terminator ready reaction mix, 1 tube M13 (-21) primer, 1 tube pGEM control DNA, protocol manual.

The dRhodamine Terminator Cycle Sequencing Kit provides optimized dRhodamine dideoxy terminator chemistries for decreased background noise in a wide array sequencing applications.

  • Increase your signal-to-noise ratio
  • Simplify workflows with easy-to-clean-up chemistries
  • Get optimized chemistry for sequencing templates with homo-polymers (AA's, TT's, etc.)
  • Obtain clean signal, even peak patterns, and minimal mobility shifts with improved dye terminators
  • Reduce spectrum overlap with narrow dye emission spectra

Specifications

Content And Storage Each kit contains 800uL Tube of dRhodamine Terminator Ready Reaction Mix, 1 Tube M13 (-21) Primer, 1 Tube pGEM Control DNA, Protocol Manual. Store kit at -15 to -25°C and avoid repeated freeze-thaw cycles.
Format 8-well Strip, 96-well Plate
For Use With (Equipment) 310 Genetic Analyzer, 3130xl Genetic Analyzer, ABI PRISM™ 377, GeneAmp 2700, GeneAmp 9600, GeneAmp 9700, GeneAmp™ 2400, 3130 Genetic Analyzer
For Use With (Application) Standard Sequencing
Includes dRhodamine Dye Terminator Ready Reaction Cycle Sequencing Kit, 100 rxns, 8-well Strips, 96-well Plate, For 310 Genetic Analyzer, 3130 Genetic Analyzer
No. of Reactions 100 Reactions
Product Type Sequencing Kit
Quantity 100 reactions
Technique Fluorescent Dye Terminator Sequencing
Template Compatibility Single Stranded DNA, Plasmid DNA (≤15Kb), PCR Amplicons

For Research Use Only. Not for use in diagnostic procedures.