missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Drebrin Recombinant Rabbit Monoclonal Antibody (23GB5145)
Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA552805
Description
Drebrin Recombinant Monoclonal Antibody for Flow, ICC/IF, Western Blot
The protein encoded by this gene is a cytoplasmic actin-binding protein thought to play a role in the process of neuronal growth. It is a member of the drebrin family of proteins that are developmentally regulated in the brain. A decrease in the amount of this protein in the brain has been implicated as a possible contributing factor in the pathogenesis of memory disturbance in Alzheimer's disease. At least two alternative splice variants encoding different protein isoforms have been described for this gene.Chemical Identifiers
| Drebrin | |
| -20°C | |
| Recombinant Monoclonal | |
| Liquid | |
| IgG | |
| Human, Mouse, Rat | |
| DBN1 | |
| D0S117E; Dbn1; developmentally regulated brain protein; developmentally-regulated brain protein; DKFZp434D064; Drba; Drebrin; drebrin 1; drebrin A; drebrin E; drebrin E2; drebrin-1; Unknown (protein for MGC:138067) | |
| DBN1 | |
| Primary | |
| Affinity chromatography |
| 23GB5145 | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q07266, Q16643, Q9QXS6 | |
| 1627, 56320, 81653 | |
| A synthesized peptide derived from human Drebrin (120-181AA). | |
| Antibody |
Specifications
| Drebrin | |
| Recombinant Monoclonal | |
| Unconjugated | |
| DBN1 | |
| D0S117E; Dbn1; developmentally regulated brain protein; developmentally-regulated brain protein; DKFZp434D064; Drba; Drebrin; drebrin 1; drebrin A; drebrin E; drebrin E2; drebrin-1; Unknown (protein for MGC:138067) | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 1627, 56320, 81653 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 23GB5145 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q07266, Q16643, Q9QXS6 | |
| DBN1 | |
| A synthesized peptide derived from human Drebrin (120-181AA). | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |