missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Drebrin Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA584319
Description
Immunogen sequence: CACASHVAKV AEFFQGVDVI VNASSVEDID AGAIGQRLSN GLARLSSPVL HRLRLREDEN AEPVGTTYQK TDAAVEMKRI NREQFWEQ Highest antigen sequence indentity to the following orthologs: Mouse - 100%, Rat - 100%.
The protein encoded by this gene is a cytoplasmic actin-binding protein thought to play a role in the process of neuronal growth. It is a member of the drebrin family of proteins that are developmentally regulated in the brain. A decrease in the amount of this protein in the brain has been implicated as a possible contributing factor in the pathogenesis of memory disturbance in Alzheimer's disease. At least two alternative splice variants encoding different protein isoforms have been described for this gene.Specifications
| Drebrin | |
| Polyclonal | |
| Unconjugated | |
| DBN1 | |
| D0S117E; Dbn1; developmentally regulated brain protein; developmentally-regulated brain protein; DKFZp434D064; Drba; Drebrin; drebrin 1; drebrin A; drebrin E; drebrin E2; drebrin-1; Unknown (protein for MGC:138067) | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 1627 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q16643 | |
| DBN1 | |
| Recombinant protein corresponding to Human DBN1. Recombinant protein control fragment (Product #RP-103282). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |