missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Drebrin Monoclonal Antibody (K56020_20H12), Invitrogen™
Description
Sequence of this protein is as follows: LELLAAYEEV IREESAADWA LYTYEDGSDD LKLAASGEGG LQELSGHFEN QKVMYGFCSV KDSQAALPKY VLINWVGEDV PDARKCACAS HVAKVAEFFQ GVDVIVNASS VEDIDAGAIG QRLSNGLARL SSPVLHRLRL REDENAEPVG TTYQKTDAAV EMKRINREQF WEQAKKEEEL RKEEERKKAL DERLRFEQER MEQERQEQEE RERRYREREQ QIEEHRRKQQ TLEAEEAKRR LKEQSIFGDH RDEEEETHMK KSESEVEEAA AIIAQRPDNP REFFKQQERV ASASAGSCDV PSPFNHRPGS HLDSHRRMAP
The protein encoded by this gene is a cytoplasmic actin-binding protein thought to play a role in the process of neuronal growth. It is a member of the drebrin family of proteins that are developmentally regulated in the brain. A decrease in the amount of this protein in the brain has been implicated as a possible contributing factor in the pathogenesis of memory disturbance in Alzheimer's disease. At least two alternative splice variants encoding different protein isoforms have been described for this gene.
Specifications
Specifications
| Antigen | Drebrin |
| Applications | Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot |
| Classification | Monoclonal |
| Clone | K56020_20H12 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide |
| Gene | DBN1 |
| Gene Accession No. | Q07266, Q16643, Q9QXS6 |
| Gene Alias | D0S117E; Dbn1; developmentally regulated brain protein; developmentally-regulated brain protein; DKFZp434D064; Drba; Drebrin; drebrin 1; drebrin A; drebrin E; drebrin E2; drebrin-1; Unknown (protein for MGC:138067) |
| Show More |