missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Drebrin Monoclonal Antibody (K56020_20H12), Invitrogen™

Catalog No. MA555629
Change view
Click to view available options
Quantity:
50 μL
200 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA555629 50 μL
MA556009 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MA555629 Supplier Invitrogen™ Supplier No. MA555629
Only null left

Mouse Monoclonal Antibody

Sequence of this protein is as follows: LELLAAYEEV IREESAADWA LYTYEDGSDD LKLAASGEGG LQELSGHFEN QKVMYGFCSV KDSQAALPKY VLINWVGEDV PDARKCACAS HVAKVAEFFQ GVDVIVNASS VEDIDAGAIG QRLSNGLARL SSPVLHRLRL REDENAEPVG TTYQKTDAAV EMKRINREQF WEQAKKEEEL RKEEERKKAL DERLRFEQER MEQERQEQEE RERRYREREQ QIEEHRRKQQ TLEAEEAKRR LKEQSIFGDH RDEEEETHMK KSESEVEEAA AIIAQRPDNP REFFKQQERV ASASAGSCDV PSPFNHRPGS HLDSHRRMAP

The protein encoded by this gene is a cytoplasmic actin-binding protein thought to play a role in the process of neuronal growth. It is a member of the drebrin family of proteins that are developmentally regulated in the brain. A decrease in the amount of this protein in the brain has been implicated as a possible contributing factor in the pathogenesis of memory disturbance in Alzheimer's disease. At least two alternative splice variants encoding different protein isoforms have been described for this gene.

Specifications

Antigen Drebrin
Applications Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Monoclonal
Clone K56020_20H12
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide
Gene DBN1
Gene Accession No. Q07266, Q16643, Q9QXS6
Gene Alias D0S117E; Dbn1; developmentally regulated brain protein; developmentally-regulated brain protein; DKFZp434D064; Drba; Drebrin; drebrin 1; drebrin A; drebrin E; drebrin E2; drebrin-1; Unknown (protein for MGC:138067)
Gene Symbols DBN1
Host Species Mouse
Immunogen Recombinant human DBN1 (immunogen range is 11-330).
Purification Method Protein A
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1627, 56320, 81653
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less