missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ DRD3 Recombinant Rabbit Monoclonal Antibody (23GB5840)

Catalog No. MA552927
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA552927 20 μL
MA552926 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MA552927 Supplier Invitrogen™ Supplier No. MA552927
Only null left

Rabbit Recombinant Monoclonal Antibody

DRD3 Recombinant Monoclonal Antibody for Flow, ICC/IF, Western Blot

This gene encodes the D3 subtype of the five (D1-D5) dopamine receptors. The activity of the D3 subtype receptor is mediated by G proteins which inhibit adenylyl cyclase. This receptor is localized to the limbic areas of the brain, which are associated with cognitive, emotional, and endocrine functions. Genetic variation in this gene may be associated with susceptibility to hereditary essential tremor 1. Alternative splicing of this gene results in transcript variants encoding different isoforms, although some variants may be subject to nonsense-mediated decay (NMD).

Specifications

Antigen DRD3
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 23GB5840
Concentration 0.333 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene DRD3
Gene Accession No. P19020, P30728, P35462
Gene Alias D(3) dopamine receptor; D3 receptor; D3DR; D3nf; D3R; Dopamine 3 receptor; Dopamine D3 receptor; dopamine receptor 3; dopamine receptor D3; dopaminergic receptor D3; Drd3; essential tremor 1; ETM1; FET1
Gene Symbols DRD3
Host Species Rabbit
Immunogen A synthesized peptide derived from human Dopamine Receptor D3 (1-50AA).
Purification Method Affinity chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 13490, 1814, 29238
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less