missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ DRAQ5™ Fluorescent Probe Solution (5 mM)

Catalog No. PI62251
Change view
Click to view available options
Quantity:
50 μL
200 μL
1 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI62251 200 μL
62252 1 mL
62254 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. PI62251 Supplier Thermo Scientific™ Supplier No. 62251

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Stain live or fixed cells with this far-red DNA stain.

Stain live or fixed cells with Thermo Scientific™ DRAQ5 Fluorescent Probe, a far-red DNA stain.

Fluorescence cell-based assays, such as flow cytometry, in-cell ELISA, fluorescence microscopy and high-content imaging require a fluorescent label to identify individual cells. When using multiple fluorescent probes to detect different cellular targets or activities, each probe must have a fluorescent spectrum different than the other probes. The blue-fluorescent DNA-binding probes, Hoechst and DAPI are frequently used; however, these probes cannot be used when UV illumination is unavailable or other blue-emitting fluorescent probes are used. Therefore, nuclear probes that emit in a color other than blue is useful for cell identification and counting, and for determining nuclear morphology and DNA content.
  • Due to its far-red excitation and emission, the DRAQ5 Fluorescent Probe can be multiplexed with many other fluorophores and is ideal for cells expressing green fluorescent protein (GFP) fusion proteins
  • Compatible with many existing protocols across a wide range of instrumentation platforms
  • Emits in the far-red region, is lipophilic and crosses cell and nuclear membranes in live and fixed cells and tissues for rapid DNA staining
  • Stain is water-soluble, supplied ready-to-use and does not require RNase, cell lysis, or a washing step, making it compatible with automation
  • Because DNA staining is stoichiometric, the DRAQ5 Fluorescent Probe can be used for DNA content analysis in cell proliferation studies

Specifications

Cell Permeability Cell-permeant
Color Red
Content And Storage Store at 4°C.
Description DRAQ5 Fluorescent Probe Solution (5 mM)
Detection Method Fluorescence
For Use With (Application) Cell Imaging
For Use With (Equipment) Fluorescence Microscope, High Content Analysis Instrument
Product Type Fluorescent Probe Solution
Dye Type Cell-Permeant
Form Liquid
Product Line Pierce
Purity or Quality Grade > 99%
Quantity 200 μL
Solubility Soluble at working concentrations in water and typical buffers
Sub Cellular Localization Nucleus, Cytosol, Cytoplasm
Excitation Wavelength 647 nm
Emission Wavelength 681
Show More Show Less
Does DRAQ5 Fluorescent Probe Solution contain DMSO?

The aqueous solution that DRAQ5 Fluorescent Probe Solution (Cat. No. 62254) is manufactured in is proprietary.

For Research Use Only. Not for use in diagnostic procedures.