missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ eBioscience™ DRAQ5™

Catalog No. 5016967
Change view
Click to view available options
Quantity:
50μL
200μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
5016967 50μL
5016968 200μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 5016967 Supplier Invitrogen™ Supplier No. 65088092

Anthraquinone dye with high affinity for double-stranded DNA. It is a membrane-permeable dye that can label live or fixed/dead cells.

  • In flow cytometry, this dye can be used to distinguish nucleated and non-nucleated cells.
  • DRAQ5™ can also be used to report nuclear DNA content for ploidy and cell cycle analysis because it binds DNA stoichiometrically. In fluorescent microscopy, it can be used as a nuclear counterstain
  • Excited by wavelengths from 488 to 647nm
  • For microscopy/imaging, 633 or 647nm sources for excitation are recommended
  • For flow cytometry, with excitation at 488nm, this dye can be detected with filter sets designed for PerCP-eFluor™ 710 or PerCP-Cy5.5 (685LP dichroic mirror and 710/50 band pass); with excitation at 633nm, it can be detected with filter sets designed for APC (660/20 band pass)
  • For cell cycle/DNA analysis applications, a higher wavelength filter is recommended, such as a 710LP or a filter set designed for PE-Cy7 (735LP dichroic mirror and 780/60 band pass), to optimize the CV for the G1 and G2/M peaks
  • Because of its broad excitation and emission properties, DRAQ5™ is not recommended in combination with other far-red fluorochromes excited by the 488 or 633nm laser lines, including PE-Cy7, PerCP-eFluor™ 710, PerCP-Cy5.5, APC, Alexa Fluor™ 647, Alexa Fluor™ 700, APC-eFluor™ 780 and equivalents (APC-Alexa Fluor™ 750 and APC-Cy7)

Specifications

Color Red
Detection Method Fluorescence
For Use With (Application) Immunohistochemistry (IHC), Flow Cytometry
For Use With (Equipment) Fluorescence Microscope
Product Type Dye
Emission 488 to 647/633 to 710
Product Line eBioscience
Quantity 50μL

For Research Use Only. Not for use in diagnostic procedures.