missing translation for 'onlineSavingsMsg'
Learn More

Dräger™ Short-Term Detector Tubes: Nitrogen Dioxide

Catalog No. 19453092
Change view
Click to view available options
Measurement Range:
0.1 to 30ppm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Measurement Range
19453092 0.1 to 30ppm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19453092 Supplier Dräger™ Supplier No. 8103631

Cost-effective and reliable way to measure toxic gases and vapors in the workplace

DragerTubes™ with the accuro™ pump deliver accurate results offering a ±10% standard deviation. This is because of the consistent volume delivered by the bellows pump design and the quality attained by individually calibrating each batch of DragerTubes, which are batch tested at regular intervals, to guarantee accuracy over the entire life of the tubes.
  • Measuring Range: 0.1 to 30ppm

Specifications

Detection Limits 0.1 to 30ppm
Components Measured Nitrogen Dioxide 0.1/a
Measurement Range 0.1 to 30ppm
Quantity 10/Pk.