missing translation for 'onlineSavingsMsg'
Learn More

Dräger™ Detector Tube - Hydrocyanic Acid

Catalog No. 19453091
Change view
Click to view available options
Components Measured:
Hydrocyanic Acid
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Components Measured
19453091 Hydrocyanic Acid
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19453091 Supplier Dräger™ Supplier No. 8103601

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

For short-term gas measurement system.

  • Sealed glass vial
  • Contains solid carrier material and chemical reagent which react to particular gas/vapour
  • Provides exact results
  • Calibration showed on in form of scale printed on tube
  • Applications in industry, fire fighting, diaster prevention, laboratory work and environmental protection

Specifications

Detection Limits 0.5 to 100ppm
Components Measured Hydrocyanic Acid
For Use With (Equipment) Draeger Accuro Pump
Type Gas Detection Tube
Material Glass
Quantity 10/Pk.
Tube Type Detector Tube