missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DR Instruments Specimen Dissecting Forceps, 6 in., Medium Points
Forceps for specimen handling in various dissection purposes.
Supplier: DR Instruments 19EMD
Description
- Crafted with surgical grade stainless-steel
- Features serrated points and knurled shanks for grip
- Length: 15.24 cm (6 in.)
Spécifications
| Dissecting Forceps | |
| Stainless Steel (Surgical Grade) | |
| 6 in. | |
| 15.24 cm |