missing translation for 'onlineSavingsMsg'
Learn More

Dräger™ Silane (PH3) XXS Replacement Sensor

Catalog No. 17980805
Change view
Click to view available options
Components Measured:
Silane
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Components Measured
17980805 Silane
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17980805 Supplier Dräger™ Supplier No. 6810886

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

XXS Replacement Sensor, for use with Pac 7000 and X-am 5000, X-am 5600 multi-gas detectors.

  • Measuring range: 0 to 20 ppm
  • Resolution: 0.01 ppm
  • Warranty: 1 year
  • Silane (Phand3)

Specifications

Components Measured Silane
For Use With (Equipment) Pac 7000, X-am 5000, X-am 5600 Multigas Detectors
Type Replacement Sensor