Learn More
Dräger™ Short-Term Detector Tubes - Phosgene
Supplier:  Dräger™ 8101521
 View more versions of this product
Catalog No. 17985288
Only null left
Description
Description
Specifications
Chemical Identifiers
3 to 15mg/L
10
Specifications
Phosgene 0.02/a
0.02 to 1ppm
10
32°F to 104°F
3 to 15mg/L
20/40
6 to 12 min.
0°C to 40°C
Product Suggestions
Product Suggestions