Learn More
Dräger™ Short-Term Detector Tubes - Ozone
Catalog No. 17985280
Change view
Click to view available options
Components Measured:
Ozone 0.05/b
Ozone 10/a
Measurement Range:
0.05 to 0.7ppm
20 to 300ppm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Components Measured Measurement Range
17985280 Ozone 0.05/b 0.05 to 0.7ppm
17985281 Ozone 10/a 20 to 300ppm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 17985280 Supplier Dräger™ Supplier No. 6733181
For making spot measurements for various gases and vapors
  • Glass vial
  • Calibrated 100mL sample of air drawn through tube with Drager Accuro™ bellows pump
  1. Extension of measuring range listed is possible for some detector tubes. Call Fisher Scientific™ at 1-800-772-6733 for more information.
  2. Safety Equipment Institute (SEI) certified.
Specifications
Components Measured Ozone 0.05/b
Measurement Range 0.05 to 0.7ppm
Quantity 10/Box