missing translation for 'onlineSavingsMsg'
Learn More

Dräger™ Short-Term Detector Tubes - Mercaptan

Catalog No. 18556G
Change view
Click to view available options
Measurement Range:
20 to 100ppm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Measurement Range
18556G 20 to 100ppm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18556G Supplier Dräger™ Supplier No. 8101871

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

These reliable, pre-calibrated detector tubes enable the rapid and precise measurement of Mercaptan gas concentrations in workplace environments and confined spaces.

The Dräger Mercaptan 20/a detector tube is designed for the rapid and precise measurement of mercaptan gas concentrations. The system uses glass tubes containing a chemical reagent that changes color in the presence of the target analyte, allowing for semi-quantitative concentration readings.

  • Gas Measured: Mercaptan
  • Standard Measurement Range: 20 to 100 ppm
  • Number of Strokes (n): 10
  • Measurement Duration: approx. 2.5 min
  • Standard Deviation: ±10 to 15 %
  • Color Change: White to yellow-brown
  • Operating Temperature: 0°C to 50°C
  • Operating Humidity: 3 to 30 mg H2O/L
  • Measurement Principle: 2 R-SH + Cu²+ → Cu(RS)² + 2 H+; Cu(RS)² + S → yellow-brown Cu-compound
  • Compatibility: Used with Dräger Accuro™ bellows pump or automatic pumps like Accuro 2000 and Quantimeter 1000
  • Cross-Sensitivity: Higher alkyl mercaptans (propyl- and butyl-mercaptan) are indicated with similar sensitivity. Measurement is not possible in the presence of hydrogen sulfide, as it is indicated with approximately double the sensitivity.

  1. Extension of measuring range listed is possible for some detector tubes. Call Fisher Scientific™ at 1-800-772-6733 for more information.
  2. Safety Equipment Institute (SEI) certified.

Specifications

Components Measured Mercaptan 20/a (Ethanethiol)
For Use With (Equipment) Accuro Drager Gas Detector Pump
Type Gas Detection Tube
Humidity Range 3 to 30mg/L
Measurement Range 20 to 100ppm
No. of Strokes 10
Quantity 10
Response Time 2.5 min.
Temperature Range (English) 32°F to 122°F
Temperature Range (Metric) 0°C to 50°C