missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dräger™ Short-Term Detector Tubes - Hydrogen Sulfide
Description
Dräger short-term detector tubes are glass vials containing a chemical reagent that provides a cost-effective and reliable method for gas measurement. The reagent changes color upon reaction, and the length of the color change directly indicates the gas concentration.
- Gas Detected: Hydrogen sulfide
- Sample Volume: 100 mL
- Measurement Time: 5 seconds to 15 minutes
- Indication Method: Reagent color change
- Compatible Pumps: Dräger Accuro™ bellows pump, Accuro 2000, or Quantimeter 1000
- Applications: Determination of concentration peaks, exposure measurement, leakage detection, and analysis of air in confined spaces such as sewers, shafts, or tanks
Specifications
Specifications
| Components Measured | Hydrogen Sulfide 1/d |
| Type | Gas Detection Tube |
| Humidity Range | 40mg/L |
| Measurement Range | 1 to 200ppm |
| No. of Strokes | 1/10 |
| Quantity | 10 |
| Response Time | 6 min. |
| Temperature Range (English) | 35.6°F to 104°F |
| Temperature Range (Metric) | 2° to 40°C |