Learn More
Dräger™ Short-Term Detector Tubes - Hydrogen Sulfide
Supplier:  Dräger™ CH29101
 View more versions of this product
Catalog No. 17985249
Only null left
Description
Description
Specifications
Chemical Identifiers
10
Specifications
Hydrogen Sulfide 100/a
100 to 2000ppm
10
32°F to 104°F
3 to 40mg/L
1
30 sec.
0°C to 40°C
Product Suggestions
Product Suggestions