Learn More
Dräger™ Short-Term Detector Tubes - Hydrogen Sulfide
Supplier:  Dräger™ CH29801
 View more versions of this product
Catalog No. 17985248
Only null left
Description
Description
Specifications
Chemical Identifiers
10
Specifications
Hydrogen Sulfide 5/b
5 to 60ppm
10
32°F to 140°F
Less than40mg/L
10
4 min.
0°C to 60°C
Product Suggestions
Product Suggestions