Learn More
Dräger™ Short-Term Detector Tubes - Hydrogen Sulfide
Supplier:  Dräger™ 6728041
 View more versions of this product
Catalog No. 17985245
Only null left
Description
Description
Specifications
Chemical Identifiers
10
Specifications
Hydrogen Sulfide 0.5/a
0.5 to 15ppm / 5 to 150ppm
10
32°F to 104°F
3 to 30mg/L
10
6 min.
0°C to 40°C
Product Suggestions
Product Suggestions