missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dräger™ Short-Term Detector Tubes - De Tube Mercaptan
Description
Dräger short-term detector tubes for the measurement of Mercaptan gas concentrations.
- Standard measuring range: 0.5 to 5 ppm
- Number of strokes (n): 20
- Measurement time: approx. 5 min.
- Standard deviation: ±10 to 15 %
- Color change: White → Yellow
- Temperature range: 10°C to 40°C
- Absolute humidity: 3 to 15 mg H2O / L
- Reaction principle: 2 R-SH + Pd2+ → Pd(RS)2 + 2 H+
- Cross sensitivity: Higher molecular weight alkyl mercaptans (e.g., propyl- and butylmercaptans) are indicated with approximately the same sensitivity.
- Interfering gases: 1,000 ppm ethylene, 2,000 ppm carbon monoxide and 200 ppm hydrogen sulfide do not affect the indication.
- Pre-layer discoloration: Hydrogen sulfide discolors the pre-layer black.
- Extension of measuring range listed is possible for some detector tubes. Call Fisher Scientific™ at 1-800-772-6733 for more information.
- Safety Equipment Institute (SEI) certified.
Specifications
Specifications
| Components Measured | Mercaptan 0.5/a |
| For Use With (Equipment) | Draeger Accuro Pump |
| Type | Gas Detection Tube |
| Humidity Range | 3 to 15mg/L |
| Measurement Range | 0.5 to 5ppm |
| No. of Strokes | 20 |
| Quantity | 10 |
| Response Time | 5 min. |
| Temperature Range (English) | 50°F to 104°F |
| Temperature Range (Metric) | 10° to 40°C |