Learn More
Dräger™ Short-Term Detector Tubes - De Tube Hydrogen
Catalog No. 17985243
Change view
Click to view available options
Components Measured:
Hydrogen Fluoride 1.5/b (gas or vapor)
Hydrogen Peroxide 0.1/a (gas or vapor)
Measurement Range:
0.1 to 3ppm
15ppm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Components Measured Measurement Range
17985243 Hydrogen Fluoride 1.5/b (gas or vapor) 15ppm
17985244 Hydrogen Peroxide 0.1/a (gas or vapor) 0.1 to 3ppm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 17985243 Supplier Dräger™ Supplier No. CH30301
  • Glass vial
  • Calibrated 100mL sample of air drawn through tube with Drager Accuro™ bellows pump
Order Info
Extension of measuring range listed is possible for some detector tubes. Call Fisher Scientific™ at 1-800-772-6733 for more information.
Specifications
Components Measured Hydrogen Fluoride 1.5/b (gas or vapor)
Measurement Range 15ppm
Quantity 10/Box