missing translation for 'onlineSavingsMsg'
Learn More

Dräger™ Short-Term Detector Tubes: Carbon Dioxide

Catalog No. 29663363
Change view
Click to view available options
Measurement Range:
0.1 to 6%
1 to 20%
100 to 3000
5 to 60%
Tube Type:
Carbon dioxide 0.1%/a2
Carbon dioxide 1%/a
Carbon dioxide 100/a
Carbon dioxide 100/a-p
Carbon dioxide 5%/A
Humidity Range:
23mg/L
30mg/L
40mg/L
50mg/L
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Measurement Range Tube Type Humidity Range
29663363 100 to 3000 Carbon dioxide 100/a-p 23mg/L
18556C 100 to 3000 Carbon dioxide 100/a 23mg/L
17985191 0.1 to 6% Carbon dioxide 0.1%/a2 30mg/L
17985194 5 to 60% Carbon dioxide 5%/A 50mg/L
17985193 1 to 20% Carbon dioxide 1%/a 40mg/L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 29663363 Supplier Dräger™ Supplier No. 6728521

Detector tubes

Glass detector tubes are filled with an inert reagent carrier material (silica gel, aluminum oxide, or aluminum silicate) impregnated with an indicating reagent. Reagent produces a colorimetric indication in the presence of a particular gas, vapor, or aerosol. Concentrations can be read directly from the color change on the tube's printed scale. Tubes are hermetically sealed. Most have a two-year shelf life (when stored at room temperature). Available in packs of ten.

Note: Some tests require a pretube or comparison tube in addition to the detector tube. These tubes are included in the box of 10, thus reducing the number of detector tubes. The actual number of detector tubes is cited in the ordering charts.

Order Info

Extension of measuring range listed is possible for some detector tubes. Call Fisher Scientific™ at 1-800-772-6733 for more information.

Specifications

Components Measured Carbon Dioxide
Humidity Range 23mg/L
Measurement Range 100 to 3000
Quantity 10
Response Time 5 min.
Temperature Range (English) 59°F to 77°F
Temperature Range (Metric) 15° to 25°C
Tube Type Carbon dioxide 100/a-p