Learn More
Dräger™ Short-Term Detector Tubes: Benzene
Detector tubes
$228.02 - $277.31
Specifications
Components Measured Benzene
View More Specs
Products 3
Catalog Number Mfr. No. Measurement Range Tube Type Humidity Range Price Quantity & Availability  
Catalog Number Mfr. No. Measurement Range Tube Type Humidity Range Price Quantity & Availability  
18556B
View Documents Promotion Details
Dräger™
8101741
15 to 420 Benzene 15/a 1 to 30mg/L
Pack of 10 Each for $228.02
Only null left
17985187
View Documents Promotion Details
Dräger™
8101231
2 to 60 Benzene 2/a 1 to 15mg/L
Pack of 10 Each for $237.70
Only null left
17985188
View Documents Promotion Details
Dräger™
6728071
5 to 50 Benzene 5/b 3 to 15mg/L
Pack of 10 Each for $277.31
Only null left
Description
Description
Glass detector tubes are filled with an inert reagent carrier material (silica gel, aluminum oxide, or aluminum silicate) impregnated with an indicating reagent. Reagent produces a colorimetric indication in the presence of a particular gas, vapor, or aerosol. Concentrations can be read directly from the color change on the tube's printed scale. Tubes are hermetically sealed. Most have a two-year shelf life (when stored at room temperature). Available in packs of ten. Note: Some tests require a pretube or comparison tube in addition to the detector tube. These tubes are included in the box of 10, thus reducing the number of detector tubes. The actual number of detector tubes is cited in the ordering charts.
Order Info
Extension of measuring range listed is possible for some detector tubes. Call Fisher Scientific™ at 1-800-772-6733 for more information.
Specifications
Specifications
Benzene