Learn More
Dräger™ Short-Term Detector Tubes: Amine Test
Catalog No. 17985185
Change view
Click to view available options
Measurement Range:
Qualitative
Catalog No. Measurement Range
17985185 Qualitative
1 options
Catalog No. 17985185 Supplier Dräger™ Supplier No. 8101061
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Detector tubes
Glass detector tubes are filled with an inert reagent carrier material (silica gel, aluminum oxide, or aluminum silicate) impregnated with an indicating reagent. Reagent produces a colorimetric indication in the presence of a particular gas, vapor, or aerosol. Concentrations can be read directly from the color change on the tube's printed scale. Tubes are hermetically sealed. Most have a two-year shelf life (when stored at room temperature). Available in packs of ten.
Note:
Some tests require a pretube or comparison tube in addition to the detector tube. These tubes are included in the box of 10, thus reducing the number of detector tubes. The actual number of detector tubes is cited in the ordering charts.
Order Info
Extension of measuring range listed is possible for some detector tubes. Call Fisher Scientific™ at 1-800-772-6733 for more information.
Specifications
Description Detector Tubes
Components Measured Basic Compounds in Air
Humidity Range 3 to 15mg/L
Measurement Range Qualitative
No. of Strokes 1
Quantity 10
Response Time 5 sec.
Temperature Range (English) 32°F to 104°F
Temperature Range (Metric) 0°C to 40°C
Tube Type Amine Test