missing translation for 'onlineSavingsMsg'
Learn More

Dräger™ Pac™ III Sensors

Catalog No. 29801486
Change view
Click to view available options
Components Measured:
Carbon Monoxide-HC (XS)
Dimethyl Sulfide
Hydrogen Sulfide (XS)
Hydrogen Sulfide-HC (XS)
Nitric Oxide
Oxygen (XS)
Phosgene (COCL2)
Measurement Range:
0 to 100
0 to 1000
0 to 10000
0 to 25%
0 to 3
0 to 40
Warranty:
1 Year
3 Years
6 Months
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Components Measured Measurement Range Warranty
29801486 Oxygen (XS) 0 to 25% 3 Years
29801484 Hydrogen Sulfide-HC (XS) 0 to 1000 1 Year
19815829 Hydrogen Sulfide (XS) 0 to 100 3 Years
19815830 Carbon Monoxide-HC (XS) 0 to 10000 1 Year
19815828 Phosgene (COCL2) 0 to 3 6 Months
19815833 Dimethyl Sulfide 0 to 40 1 Year
19815836 Nitric Oxide 0 to 100 1 Year
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 29801486 Supplier Dräger™ Supplier No. 6809130

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Provide fast and accurate responses to hazardous gas levels

  • For use with Pac™ III single gas monitors

Specifications

Components Measured Oxygen (XS)
Humidity Range 10 to 95%
Measurement Range 0 to 25%
Pressure Range 700 to 1300mbar
Resolution 0.1%
Temperature Range (English) -40°F to +122°F
Temperature Range (Metric) -40° to +50°C
Warranty 3 Years