Learn More
Dräger™ Pac™ III Sensors
Provide fast and accurate responses to hazardous gas levels
Supplier:  Dräger™ 6809200
 View more versions of this product
Catalog No. 19815833
Only null left
Description
Description
  • For use with Pac™ III single gas monitors
Specifications
Chemical Identifiers
700 to 1300mbar
Specifications
Dimethyl Sulfide
0 to 40
0.1
-40° to +50°C
10 to 90%
700 to 1300mbar
-40°F to +122°F
1 Year
Product Suggestions
Product Suggestions