Learn More
Dräger™ Individual Haz Mat Simultaneous Test Sets
Catalog No. 18556S
Change view
Click to view available options
Type:
Simultaneous Test Set I
Simultaneous Test Set II
Simultaneous Test Set III
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
18556S Simultaneous Test Set II
18556R Simultaneous Test Set I
18556T Simultaneous Test Set III
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
No other type of tube should be substituted with these kits.
Catalog No. 18556S Supplier Dräger™ Supplier No. 8101736
No other type of tube should be substituted with these kits.
Requires
Require adapter and a Dräger pump (sold separately) for the semi-quantitative determination of inorganic gases and organic vapors.
For simultaneous testing of five different gases
Test Set I. Includes detector tubes for hydrogen chloride, hydrogen cyanide, carbon monoxide, ammonia, and nitrous fumes.
Test Set II. Includes detector tubes for sulfur dioxide, chlorine, hydrogen sulfide, carbon dioxide, and phosgene.
Test Set III. Includes detector tubes for alcohols, aliphatic hydrocarbons, aromatics, chlorinated hydrocarbons, and ketones.
Order Info
Packs of 10 test sets.
Specifications
Type Simultaneous Test Set II
For Use With (Application) For simultaneous testing of five different gases
Includes Tests for sulfur dioxide, chlorine, hydrogen sulfide, carbon dioxide, and phosgene