missing translation for 'onlineSavingsMsg'
Learn More

Dräger™ Hydrogen Sulfide (H2S) XXS Replacement Sensor

Catalog No. 196810883
Change view
Click to view available options
Components Measured:
Hydrogen Sulfide
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Components Measured
196810883 Hydrogen Sulfide
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 196810883 Supplier Dräger™ Supplier No. 6810883

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

XXS Replacement Sensor, for use with Pac 7000, X-am 2000, X-am 5000 multi-gas detectors.

  • Measuring range: 0 to 100 ppm
  • Resolution: 1 ppm
  • Warranty: 2 years
  • Hydrogen Sulfide (H2S)

Specifications

Components Measured Hydrogen Sulfide
For Use With (Equipment) Pac 7000, X-am 5000, X-am 5600 Multigas Detectors
Type Replacement Sensor