missing translation for 'onlineSavingsMsg'
Learn More

Dräger™ Carbon Dioxide (CO2) XXS Replacement Sensor

Catalog No. 17980808
Change view
Click to view available options
Components Measured:
Carbon Dioxide
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Components Measured
17980808 Carbon Dioxide
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17980808 Supplier Dräger™ Supplier No. 6810889

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

XXS Replacement Sensor, for use with Pac 7000, X-am 5000 multi-gas detectors.

  • Measuring range: 0 to 5% vol.
  • Resolution: 0.1% vol.
  • Carbon Dioxide (CO2)

Specifications

Components Measured Carbon Dioxide
For Use With (Equipment) Pac 7000, X-am 5000 Multigas Detectors
Type Replacement Sensor